Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.93658

Supelco

Barium chloride dihydrate

EMPLURA®

Synonym(s):

Barium chloride dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
BaCl2 · 2H2O
CAS Number:
Molecular Weight:
244.26
MDL number:
UNSPSC Code:
12352302

product line

EMPLURA®

storage temp.

room temp

InChI

1S/Ba.2ClH.2H2O/h;2*1H;2*1H2/q+2;;;;/p-2

InChI key

PWHCIQQGOQTFAE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Eye Irrit. 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service