Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.93500

Supelco

Ammonia solution

EMPLURA®

Synonym(s):

Ammonium hydroxide solution, Ammonia aqueous, Ammonia water

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4OH
CAS Number:
Molecular Weight:
35.05
Beilstein:
3587154
MDL number:
UNSPSC Code:
12352106

grade

laboratory grade

vapor pressure

483 hPa ( 20 °C)

product line

EMPLURA®

form

liquid

expl. lim.

27 %

pH

(20 °C, strongly alkaline)

bp

37.7 °C/1,013 hPa

mp

-57.5 °C (-71.5 °F)

solubility

water: soluble

density

0.9 g/mL at 25 °C (lit.)

storage temp.

20-25°C

SMILES string

[NH4+].[OH-]

InChI

1S/H3N.H2O/h1H3;1H2

InChI key

VHUUQVKOLVNVRT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

1.93500.0521: 500ML, Minimum order quantity: 20
1.93500.2521: 2.5 L, Minimum order quantity: 4
1.93500.5021: 5 L Minimum Order Quantity: 4
1.93500.9026: 25 kg, Minimum order quantity: 1
1.93500.9121: 105 L, Minimum order quantity: 1

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service