Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.93406

Supelco

Nitric acid

~70%, EMPLURA®

Synonym(s):

Nitric acid solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HNO3
CAS Number:
Molecular Weight:
63.01
MDL number:
UNSPSC Code:
12352106

product line

EMPLURA®

Assay

~70%

storage temp.

20-25°C

SMILES string

O[N+]([O-])=O

InChI

1S/HNO3/c2-1(3)4/h(H,2,3,4)

InChI key

GRYLNZFGIOXLOG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

1.93406.0521: 500ML, Minimum order quantity: 16
1.93406.2521: 2.5L, Minimum order quantity: 4
1.93406.5021: 5 L Minimum Order Quantity: 2

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Eye Dam. 1 - Met. Corr. 1 - Ox. Liq. 3 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service