Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.93282

Supelco

Aluminium ammonium sulfate dodecahydrate

for analysis EMPARTA® ACS

Synonym(s):

Aluminium ammonium sulfate dodecahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4Al(SO4)2 · 12 H2O
CAS Number:
Molecular Weight:
453.33
MDL number:
UNSPSC Code:
12352302

grade

ACS reagent

storage temp.

room temp

InChI

1S/Al.H3N.2H2O4S.12H2O/c;;2*1-5(2,3)4;;;;;;;;;;;;/h;1H3;2*(H2,1,2,3,4);12*1H2/q+3;;;;;;;;;;;;;;;/p-3

InChI key

WZUKKIPWIPZMAS-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Packaging

1.93282.0521: 500 g, Minimum order quantity: 20
1.93282.5021: 5 kg, Minimum order quantity: 4

Legal Information

EMPARTA is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service