1.93226
Potassium sodium tartrate tetrahydrate
for analysis EMPARTA® ACS
Synonym(s):
Potassium sodium tartrate tetrahydrate
About This Item
Recommended Products
grade
ACS reagent
storage temp.
room temp
SMILES string
[K+].[Na+].[O-]C(=O)[C@H](O)[C@@H](O)C(=O)[O-].O.O.O.O
InChI
1S/C4H6O6.K.Na.4H2O/c5-1(3(7)8)2(6)4(9)10;;;;;;/h1-2,5-6H,(H,7,8)(H,9,10);;;4*1H2/q;2*+1;;;;/p-2/t1-,2-;;;;;;/m1....../s1
InChI key
VZOPRCCTKLAGPN-ZFJVMAEJSA-L
Looking for similar products? Visit Product Comparison Guide
Packaging
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service