Skip to Content
MilliporeSigma
All Photos(1)

Documents

189482

Sigma-Aldrich

Autocamtide-2 Related Inhibitory Peptide, Myristoylated

The Autocamtide-2 Related Inhibitory Peptide, Myristoylated controls the biological activity of calmodulin-dependent protein kinase II. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Autocamtide-2 Related Inhibitory Peptide, Myristoylated, Myristoylated AIP, Myr-N-KKALRRQGAVDAL-OH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C78H142N22O20
Molecular Weight:
1708.10
UNSPSC Code:
12352202
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

solubility

water: 1 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

This peptide corresponds to AIP (Cat. No. 189480) which has been myristoylated at the N-terminus, thus enhancing its cell-permeability.
This peptide corresponds to AIP (Cat. No. 189480) which has been myristoylated at the N-terminus, enhancing its cell-permeability.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Calmodulin-Dependent Protein Kinase (CaM Kinase)-2
Product does not compete with ATP.
Reversible: no
Target IC50: 40 nM against cam Kinase-2

Warning

Toxicity: Standard Handling (A)

Sequence

Myr-N-Lys-Lys-Ala-Leu-Arg-Arg-Gln-Glu-Ala-Val-Asp-Ala-Leu-OH

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Gailly, P., et al. 1998. Cell Calcium 24, 293.
Ishida, A., and Fujisawa, H. 1995. J. Biol. Chem.270, 2163.
Ishida, A., et al. 1995. Biochem. Biophys. Res. Commun.212, 806.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service