Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

189404

Sigma-Aldrich

Aurora Kinase Inhibitor II

The Aurora Kinase Inhibitor II, also referenced under CAS 331770-21-9, controls the biological activity of Aurora Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Aurora Kinase Inhibitor II, 4-(4ʹ-Benzamidoanilino)-6,7-dimethoxyquinazoline, Aurora Kinase Inhibitor II

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H20N4O3
CAS Number:
Molecular Weight:
400.43
UNSPSC Code:
12352200

Quality Level

Assay

≥90% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C23H20N4O3/c1-29-20-12-18-19(13-21(20)30-2)24-14-25-22(18)26-16-8-10-17(11-9-16)27-23(28)15-6-4-3-5-7-15/h3-14H,1-2H3,(H,27,28)(H,24,25,26)

InChI key

IMYVCWQAHSYYOO-UHFFFAOYSA-N

General description

A cell-permeable anilinoquinazoline compound that acts as a potent, selective, and ATP-competitive inhibitor of Aurora kinases (IC50 = 310 nM and 240 nM for Aurora A and B, respectively, 1.25 µM in MCF7 cells). Due to its limited aqueous solubility and high serum binding property, it is recommended for in vitro, but not in vivo, use.
A cell-permeable, potent, selective, and ATP-competitive inhibitor of Aurora kinases (IC50 = 310 nM and 240 nM for Aurora A and B, respectively, 1.25 µM in MCF7 cells). Due to its limited aqueous solubility and high serum binding property, it is recommended for in vitro, but not in vivo, use.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Aurora Kinase
Product competes with ATP.
Reversible: no
Target IC50: 310 nM and 240 nM for Aurora A and B, respectively, 1.25 µM in MCF7 cells

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Heron, N.M., et al. 2006. Bioorg. Med. Chem. Lett.16, 1320.
Mortlock, A.A., et al. 2005. Curr. Top. Med. Chem.5, 807.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service