Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

182800

Sigma-Aldrich

Ascomycin, Streptomyces hygroscopicus

An ethyl analog of FK506 with similar immunosuppressant properties.

Synonym(s):

Ascomycin, Streptomyces hygroscopicus, Immunomycin, FR-900520

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C43H69NO12
CAS Number:
Molecular Weight:
792.01
UNSPSC Code:
12352200

Quality Level

Assay

≥90% (HPLC)

form

crystalline solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

DMSO: 1 mg/mL
ethanol: 1 mg/mL

shipped in

ambient

storage temp.

−20°C

InChI

1S/C43H69NO12/c1-10-30-18-24(2)17-25(3)19-36(53-8)39-37(54-9)21-27(5)43(51,56-39)40(48)41(49)44-16-12-11-13-31(44)42(50)55-38(28(6)33(46)23-34(30)47)26(4)20-29-14-15-32(45)35(22-29)52-7/h18,20,25,27-33,35-39,45-46,51H,10-17,19,21-23H2,1-9H3/b24-18+,26-20+/t25-,27+,28+,29+,30+,31-,32-,33-,35+,36-,37-,38+,39+,43+/m0/s1

InChI key

ZDQSOHOQTUFQEM-BFFCMSJVSA-N

General description

An ethyl analog of FK506 with similar immunosuppressant properties. Acts by binding to immunophilins. Inhibits the synthesis of Th-1 and Th-2 type cytokines in target cells.
Ethyl analog of FK-506 that possesses the same potent immunosuppressive properties as the parent compound. Inhibition of the Ca2+/calmodulin activated phosphatase calcineurin (IC50 = 10 nM) by the ascomycin/FK-506 binding protein complex is believed to be important in the mechanism of immunosuppression. Acts by binding immumophilins.

Biochem/physiol Actions

Cell permeable: no
Fully active via yeast rotamase (FK 506 binding protein) inhibition
Primary Target
Immunophilins
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Ethanol stock solutions are stable for up to 3 days at -20°C. Anhydrous DMSO stock solutions are stable for up to 1 month -20°C.

Other Notes

Paul, C., et al. 2000. Expert Opin. Invest. Drugs9, 69.
Kawai, M., et al. 1993. FEBS Lett.316, 107.
Pilot-Matias, T.J., et al. 1993. Gene128, 219.
Shafiee, A., et al. 1993. J. Antibiot.46, 1397.
Petros, A.M., et al. 1992. J. Med. Chem. 35, 2467.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service