Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

17851-M

Millipore

Aquacide II

Synonym(s):

Aquacide II

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
$153.00
5 KG
$614.00

$153.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 KG
$153.00
5 KG
$614.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
41106500
NACRES:
NA.21

$153.00


Please contact Customer Service for Availability

Request a Bulk Order

description

RTECS - FJ5950000
Merck USA index - 14, 1829

Quality Level

100
200

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

storage temp.

10-30°C

SMILES string

[Na].OC(C(O)C(O)C=O)C(O)CO.OC(=O)C

InChI

1S/C6H12O6.C2H4O2.Na/c7-1-3(9)5(11)6(12)4(10)2-8;1-2(3)4;/h1,3-6,8-12H,2H2;1H3,(H,3,4);

InChI key

DPXJVFZANSGRMM-UHFFFAOYSA-N

General description

Aquacides are useful for concentration of proteins and other macromolecules and can be used for rapid removal of water from dialysis tubing by using either a 2% syrup or the Aquacide solid. Aquacides neither denature proteins nor change the pH during concentration. At room temperature 1 g of Aquacide absorbs about 5 ml of water from solution in one hour. Sodium salt of carboxymethylcellulose. Has extremely high viscosity. Contains an average of 0.85 carboxymethyl groups per anhydroglucose unit.
Sodium salt of carboxymethylcellulose. Has extremely high viscosity. Contains an average of 0.85 carboxymethyl groups per anhydroglucose unit.

Warning

Toxicity: Standard Handling (A)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service