Skip to Content
MilliporeSigma
All Photos(1)

Documents

176872

Sigma-Aldrich

Angiotensin Converting Enzyme-2, His•Tag®, Human, Recombinant, NSO Cells

Synonym(s):

ACE Homolog, ACEH, ACE-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

recombinant

expressed in NSO cells

Quality Level

Assay

≥90% (SDS-PAGE)

form

liquid

specific activity

≥0.8 units/μg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

impurities

≤1.0 EU/μg Endotoxins (EU/μg ACE-2)

shipped in

wet ice

storage temp.

−70°C

General description

Recombinant, human ACE-2 ectodomain (amino acids 1-740) fused at the C-terminus to a His•Tag sequence and expressed in NSO cells. ACE-2 cleaves angiotensins I and II as a carboxypeptidase and plays an important role in regulating cardiovascular and renal functions. Native ACE-2 is an integral membrane zinc metalloprotease whose activity is inhibited by EDTA. By SDS-PAGE, the apparent molecular mass of the glycosylated protein is ~120 kDa under either reducing or non-reducing conditions.
Recombinant, human ACE-2 ectodomain (amino acids 1-740) fused at the C-terminus to a His•Tag sequence and expressed in NSO cells. Native ACE-2 is an integral membrane zinc metalloprotease that can be inhibited by EDTA. ACE-2 cleaves angiotensins I and II. Plays an important role in regulating cardiovascular and renal functions. By SDS-PAGE, the apparent molecular mass of the glycosylated protein is ~120 kDa under either reducing or non-reducing condition.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will cleave 1 nmol MCA-YVADAPK(DNP)-OH per min at 25°C, pH 7.5.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Tikellis, C., et al. 2003. Hypertension41, 390.
Tipnis, S.R., et al. 2000. J. Biol. Chem.275, 33238.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
HIS TAG is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service