Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.751188

Millipore

Prosep® Ultra Plus

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151816
eCl@ss:
32011308

ligand

recombinant protein A

Quality Level

manufacturer/tradename

Prosep®

parameter

<800 cm/hr mobile phase velocity

bed H

300 mm

matrix

controlled pore glass (CPG) base material

particle size

60 μm

pH

1.5 – 8.5

capacity

>50 mg/mL binding capacity(dynamic; typically; 10% breakthrough at 3–6 min residence time)
≥67 mg/mL binding capacity (hIgG)(static; typically)

Looking for similar products? Visit Product Comparison Guide

Packaging

Bottle

Storage and Stability

2–8 °C, plus bacteriostat (recommended for long-term storage)

Legal Information

PROSEP is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service