Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.73650

Supelco

Empty cells

Sign Into View Organizational & Contract Pricing

Select a Size

12 UNITS
$371.00

$371.00


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
12 UNITS
$371.00

About This Item

UNSPSC Code:
41121813
NACRES:
NB.11

$371.00


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information

material

optical glass vial

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Move 100
for use with Spectroquant® Move DC

detection method

photometric

storage temp.

2-30°C

General description

with screw caps 24 mm, Spectroquant®

These empty cells are suitable for the measurement using the photometers with a cell holder for cells with 24 mm. The cells are equipped with screw caps and are delivered in a plastic box, protected from scratching.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Spectrophotometric determination of bromine with dipropyl-p-phenylenediamine (DPD) in drinking water, swimming-pool water, wastewater, and disinfection solutions in a range of 0.020 – 10.00 mg/L.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service