Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.73500

Supelco

Screw caps

for TOC digestion, Spectroquant®

Sign Into View Organizational & Contract Pricing

Select a Size

6 UNITS
$180.00

$180.00


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
6 UNITS
$180.00

About This Item

UNSPSC Code:
41116116
NACRES:
NB.11

$180.00


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information

product line

Spectroquant®

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® TOC Cell Test

storage temp.

2-30°C

General description

These screw caps are a necessary equipment for the TOC digestion with our Spectroquant® TOC test kits 114878 and 114879. After filling the Sample into the cells they are closed with the metal caps. Then the cells are tuned upside down and placed in a heating block for the digestion of TOC to CO2. For more information please refer to the respective test instructions.

Application


  • Studies of Labware Contamination during Lipid Extraction in Mass Spectrometry-Based Lipidome Analysis.: This research explores the impact of using screw caps in lipid extraction processes for mass spectrometry, highlighting potential contamination risks and suggesting methodologies for minimizing such risks in analytical chemistry settings (Canez et al., 2024).

  • An improved sensitive assay for simultaneous determination of plasma haloperidol and reduced haloperidol levels by liquid chromatography using a coulometric detector.: Although an older study, this paper details a methodology that uses screw caps in the preparation phase of samples for high-precision liquid chromatography, relevant for chemical synthesis and analytical chemistry (Hariharan et al., 1989).

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Are these caps reusable?

    1 answer
    1. The stainless steel screw caps in this product are suitable for multiple uses. It is recommended to clean them thoroughly between uses to prevent any cross-contamination.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service