Skip to Content
MilliporeSigma
All Photos(1)

Documents

171259

Sigma-Aldrich

AMPK Activator III, DHPO

The AMPK Activator III, DHPO modulates the biological activity of AMPK. This small molecule/inhibitor is primarily used for Activators/Inducers applications.

Synonym(s):

AMPK Activator III, DHPO, 1-oxo-2-(2H-pyrrolium-1-yl)-1H-inden-3-olate, AMPK Signaling Activator III

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H9NO2
Molecular Weight:
211.22
UNSPSC Code:
12352200

Quality Level

Assay

>95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

orange

solubility

DMSO: 10 mg/mL

shipped in

wet ice

storage temp.

−20°C

General description

A cell-permeable AMPK activator (20 mg/kg/d i.p. for 21d) that has been shown to attenuate fasting blood glucose, improve glucose disposal, and correct dyslipidemia in genetic (leptin deficient, ob/ob) and dietary (high-fat-fed) mouse models of insulin resistance over controls. IP injections for 14 days increases skeletal muscle glucose uptake by > ~30% and ~70% compared to vehicle-treated lean and obese mice, respectively. Treatment with 100 µM of DHPO results in a small but significant AMPK (Thr172) phosphorylation, and a concentration dependent increase in glucose uptake with a 2-fold increase over controls, in cultured myotubes. Furthermore, 20 nM Compound C demonstrates a 58% and 41% inhibition of DHPO-stimulated glucose up-take at concentrations of 50 µM and 100 µM DHPO, respectively.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Unstable in solution; reconstitute just prior to use.

Other Notes

Kandadi, M.R., et al. 2009. Biochem. Pharmacol.79, 623.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service