Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

171254

Millipore

Ampicillin, Sodium Salt

Ampicillin salt is an antibiotic that inhibits bacterial cell-wall synthesis. Active against Gram-negative bacteria. Inactivated by lactamases. Suitable for use in research that uses ampicillin-resistant plasmids.

Synonym(s):

Ampicillin, Sodium Salt, 6-[D(–)-α-Aminophenylacetamido]penicillanic Acid, Na, Alpha-Aminobenzyl Penicillin, Ampicillin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H18N3O4S · Na
CAS Number:
Molecular Weight:
371.39
MDL number:
UNSPSC Code:
41116107

Quality Level

Assay

≥90% (anhydrous)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

water: 1 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[Na+].S1[C@H]2N([C@H](C1(C)C)C(=O)[O-])C(=O)[C@H]2NC(=O)[C@H](N)c3ccccc3

InChI

1S/C16H19N3O4S.Na/c1-16(2)11(15(22)23)19-13(21)10(14(19)24-16)18-12(20)9(17)8-6-4-3-5-7-8;/h3-7,9-11,14H,17H2,1-2H3,(H,18,20)(H,22,23);/q;+1/p-1/t9-,10-,11+,14-;/m1./s1

InChI key

KLOHDWPABZXLGI-YWUHCJSESA-M

General description

Kills growing cells by interfering with the terminal reaction in bacterial wall synthesis. Active against Gram-negative bacteria. Inactivated by lactamases. Suitable for use in research that uses ampicillin-resistant plasmids.

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Aqueous stock solutions are stable for up to 1 month at -20°C. Product is heat-resistant and stable between pH 2 and 11.

Other Notes

Nakajima, M., et al. 1994. Neurosci. Lett.176, 161.
Wu, M.H., and Yung, B.Y. 1994. Eur. J. Pharmacol.270, 203.
Betzel, C., et al. 1993. Biochim. Biophys. Acta1161, 47.
Yung, B.Y., et al. 1992. Int. J. Cancer52, 317.
Martin, S.J., et al. 1990. J. Immunol.145, 1859.
Yung, B.Y., et al. 1990. Cancer Res.50, 5987.
White, R.J., and Phillips, D.R. 1985. Biochemistry27, 9122.
Madharavao, M., et al. 1978. J. Med. Chem. 21, 958.
Sengupta, S.K., et al. 1975. J. Med. Chem. 18, 1175.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1A - Skin Sens. 1A

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service