Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

17-10499-2

Sigma-Aldrich

Magna MeRIP m6A Assay, -20C

Synonym(s):

Methylated RNA Immunoprecipitation kit, RNA detection

Sign Into View Organizational & Contract Pricing

Select a Size

17-10499-2
$749.00

$749.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
17-10499-2
$749.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.84

$749.00


Please contact Customer Service for Availability

Request a Bulk Order

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. 17-10499-2, how many reactions in this kit

    1 answer
    1. The Magna MeRIP M6A Kit provides sufficient reagents for 10 MeRIP reactions.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service