Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

17-10388

Sigma-Aldrich

Di-Ubiquitin (Lys63-linked) Recombinant Protein

Native K63 linked Di-Ubiquitin which can be used as a substrate for proteases that cleave the isopeptide linkage between two ubiquitin molecules.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.52

Quality Level

Assay

>95% (reversed phase HPLC)

mol wt

Mw ~17 kDa

manufacturer/tradename

Chemicon®

shipped in

dry ice

General description

ANTIBODY AVAILABLE!
Anti-Ubiquitin, Lys63-Specific, clone Apu3 (catalogue # 05-1308)

This product is a native K63 linked Di-Ubiquitin which can be used as a substrate for proteases that cleave the isopeptide linkage between two ubiquitin molecules. It can also be used to investigate mechanism of binding and recognition by proteins that contain ubiquitin-associated domains or ubiquitin-interacting motifs (UIMs).
This product is formed by chemical ligation.
Polyubiquitin chains linked through the Lys48 residue of ubiquitin are most commonly associated with proteins targeted for proteosomal degradation.

In contrast, polyubiquitin chains linked through the Lys63 residue of ubiquitin (Ub) are associated with nonproteolytic functions such as signal transduction. Lys63-linked polyUb chains connect components of NFκB signaling in a highly regulated manner, and genetic evidence indicates the involvement of Lys63-linked Ub chains in stress response and DNA repair. Research in yeast has suggested Lys63-linked Ub chains stimulate endocytosis.

Application

LC-MS Analysis
Mobile phase A = 1% CH3CN, 0.1% formic acid in water (MilliQ) and B = 1% water (milliQ) and 0.1% formic acid in
CH3CN. Phenomenex Kinetex C18, (2.1×50 mm, 2.6 μM); flow rate = 0.6 mL/min, runtime = 6 min, column T = 40°C.
Gradient: 5% to 95% B over 3.5 min. (See Figure 2)

Components

50 µg of Di-Ubiquitin (Lys63-linked) reconstituted in 2.5 µL DMSO and 56 µL of 0.02 M TRIS, 0.25 M NaCl, pH 7.5 to a final concentration of 50 µM.

Quality

Western Blot Analysis:
0.5-2 μg/mL of anti-Ubiquitin, Lys63-Specific, clone Apu2 (Cat. # 05-1308) detected Lys63-linked Ubiquitin from 1 µg of Di-Ubiquitin (Lys63-linked) Recombinant Protein (Cat. # 17-10388). (See Figure 1)

Storage and Stability

Store at -80°C. Avoid freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F - (refers to pure substance)

Flash Point(C)

87 °C - (refers to pure substance)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service