Skip to Content
MilliporeSigma
All Photos(3)

Documents

17-10077

Sigma-Aldrich

Magna ChIP® HT96 Chromatin Immunoprecipitation Kit

The Magna ChIP HT96 kit allows the performance of chromatin Immunoprecipitation in a 96-well plate-based format.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.41

Quality Level

manufacturer/tradename

Magna ChIP®

technique(s)

immunoprecipitation (IP): suitable

shipped in

dry ice

General description

The Magna ChIP HT96 kit allows the performance of chromatin Immunoprecipitation in a 96-well plate-based format. This reliable streamlined approach works for both cells and tissues and can be performed using either a multi-channel pipette or an automated liquid handling instrument.Each kit includes a complete set of validated, quality controlled reagents, plus a detailed protocol.

Features & Benefits
  • Complete set of materials for up to 96 ChIP reactions
  • Protein A+G bead blend for ChIP with a broader range of antibodies than A or G alone
  • Low Chromatin requirements: 10, 000 to 100, 000 cells per reaction
  • Optimized streamlined protocol with only a single buffer for sonication, IP, or wash; and protocols for automated liquid handling systems
  • Protocols for using cells or tissues
  • Direct analysis of DNA without additional clean-up steps
  • Compatible with ChIPAb+ validated antibody and primer sets

Application

Research Category
Epigenetics & Nuclear Function
Used to detect/quantify: Protein A+G

Packaging

Kit capacity: 96 chromatin immunoprecipitation assays

Components

Magna ChIP protein A/G magnetic beads

HT96 Nuclei Isolation Buffer

HT96 ChIP Buffer (Sonication/ChIP/Wash)

Low Stringency IP Wash Buffer

HT96 ChIP Elution Buffer

Proteinase K Solution

Protease Inhibitor Cocktail III

10X Glycine

10X PBS

96 Well ChIP Plate

96 Well Thermal Plate

Plate Seal

Strip Caps

Physical form

Two boxes containing all necessary reagents to perform 96 chromatin immunoprecipitation reactions in a 96 well plate format. Supplied buffers are sufficient to generate 24 individual chromatin preparation reactions using 15 cm plates of cultured cells or tissue samples.

Storage and Stability

Upon receipt, store components at the temperatures indicated on the labels. Kit components are stable for 1 year from date of shipment when stored as directed.

Legal Information

MAGNA CHIP is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10-13 - German Storage Class 10 to 13

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Chromatin Immunoprecipitation qPCR for studying gene regulation across conditions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service