Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.70355

Supelco

Sulfur ICP standard

traceable to SRM from NIST H₂SO₄ in H₂O 1000 mg/l S Certipur®

Synonym(s):

Sulfur Calibration Standard for ICP

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$144.00

$144.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
$144.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$144.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

product line

Titrisol®

form

concentrate

concentration

1000 mg/L S in water

technique(s)

ICP: suitable

pH

1.5 (20 °C in H2O)

density

1.000 g/cm3 at 20 °C

application(s)

industrial qc
pharmaceutical

format

single component solution

storage temp.

15-25°C

Related Categories

General description

This certified reference material (CRM) is produced in accordance with ISO 17034 and characterized in accordance with ISO/IEC 17025. This CRM is traceable to SI unit kg and measured against primary material from a National Metrology Institute (NMI), e.g. NIST.
Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.
Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty, date of expiry, and detailed information about trace impurities.

Application

Sulfur ICP standard can be used as a calibration standard in the determination of sulfur in amino acids by inductively coupled plasma (ICP) technique.[1]

Analysis Note

1000 mg/l S in water, prepared with high-purity H2SO4, and water

Other Notes

For a complete product listing of our Certipur® range of CRMs for ICP and AAS, technical information, and example certificates please visit our ICP & AAS standards website

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany
Titrisol is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In vivo synthesized 34 S enriched amino acid standards for species specific isotope dilution of proteins
G Hermann, et al.
Journal of Analytical Atomic Spectrometry, 31, 1830-1835 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service