Key Documents
1.67207
Supelco
Reducer (PTFE) from S40 to S38
Select a Size
1 UNIT
$80.00
$80.00
Please contact Customer Service for Availability
Request a Bulk Order
Select a Size
Change View
1 UNIT
$80.00
About This Item
UNSPSC Code:
41122409
NACRES:
NB.51
$80.00
Please contact Customer Service for Availability
Request a Bulk Order
storage temp.
no temp limit
General description
Special S40 bottle adapters made of PTFE provide all possible connection options for safe and reliable daily solvents handling. This adapter is equipped with S40 inner thread and GL45 outer thread.