Skip to Content
MilliporeSigma
All Photos(1)

Documents

16-663

Sigma-Aldrich

Magna ChIP® Protein A+G Magnetic Beads

provides a rapid, reproducible and efficient collection of immunocomplexes for ChIP and RIP assays

Synonym(s):

ChIP magnetic A+G beads, ChIP magnetic A/G beads, ChIP magnetic beads

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105501
NACRES:
NA.84

Quality Level

packaging

pkg of 1 mL

manufacturer/tradename

Magna ChIP®

storage condition

do not freeze

particle size

~3 μm

shipped in

wet ice

storage temp.

2-8°C

General description

Chromatin immunoprecipitation (ChIP) has been widely adapted for the study of gene-specific and genome-wide distribution of specific DNA and RNA-binding proteins or protein modifications. Similar to standard protein immunoprecipitation assays, ChIP involves isolation of immunocomplexes using a solid medium, such as agarose or magnetic beads, coupled to either IgG binding recombinant protein A or protein G. Experiments comparing protein A vs. protein G vs. protein A/G magnetic bead blends revealed that a mixture of protein A and G beads worked well with a wide variety of antibody isotypes. The use of Protein A/G bead blends eliminated the need to consider which beads or kit to use in order match a particular antibody/bead binding affinity combination. In addition to simplifying the procedure, comparing the use of either protein A or protein G alone, protein A/G magnetic bead blends improved signal-to-noise ratios without decreasing the recovery of input chromatin in ChIP assays.

Application

This blend of protein A+G magnetic beads allows for the use of a wider range of antibodies than A or G alone & provides a rapid, reproducible & efficient collection of immunocomplexes for chromatin immunoprecipitations (ChIP) and RNA immunoprecipitations (RIP) assays.

Use 20 µL of bead suspension per ChIP application. Includes sufficient reagents for 50 precipitation reactions. Disperse beads thoroughly before pipetting by rapid vortex.

Used to detect/quantify: Protein A+G

Quality

Routinely evaluated by Chromatin immunoprecipitation (ChIP) using HeLa nuclear extracts and the Magna ChIP® A Kit (Cat. #17-610).

Physical form

Liquid suspension. Supplied as magnetic bead slurry in phosphate buffered saline, pH 7.4, containing 0.01% Tween®-20 and 0.09% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of shipment. Do Not Freeze.

Legal Information

MAGNA CHIP is a registered trademark of Merck KGaA, Darmstadt, Germany
TWEEN is a registered trademark of Croda International PLC

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service