Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.66552

Millipore

Tryptic Soy Agar

with Lecithin and Tween®, ICR+ Contact plate, irradiated, triple packed, for environmental monitoring (Isolator and Clean room)

Synonym(s):

TSA, Soybean Casein Digest Agar, ICR plus Contact plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106231
Pricing and availability is not currently available.

material

polystyrene plate

Quality Level

Agency

EP
USP

sterility

sterile; γ-irradiated

form

plate

feature

2D bar code
ready-to-use

packaging

pkg of (20/200 Plates per box)

D

55 mm

pH

(7.3 (in H2O))

application(s)

microbiology
pharmaceutical
surface monitoring

storage temp.

15-25°C

suitability

bacteria
fungi

General description

Tryptic Soy Agar – ICR plus contact plate is designed for the determination of the total microbial aerobic and anaerobic count on dry, sanitized surfaces and personnel in Clean rooms and Isolators. It is a complex medium for cultivation and isolation of a wide range of bacteria, yeasts and molds. The formulation of the Soybean Casein-Digest medium contains a combination of casein and soy peptones supplying organic nitrogen, particularly amino acids and longer-chained peptides. The medium is supplemented with Lecithin, Polysorbate 80 (Tween®) and pyruvate to inactivate potential residues of disinfectants on dry surfaces. In addition, the plate model (plus or “+”) is supplied with a lockable lid for safe transport of the samples without the risk of losing the lid. For anaerobic incubation the lid needs to be turned into the vent position.

Application

Tryptic Soy Agar (TSA, Soybean Casein Digest Agar) is a complex medium for cultivation and isolation of a wide range of bacteria, yeasts and molds. The media finds its application in environmental monitoring by determining the total microbial aerobic and anaerobic count on dry, sanitized surfaces and personnel in Isolators and Clean Rooms

Packaging

1665520020-CN: 20 Plates per box
1665520200-CN: 200 Plates per box

Legal Information

TWEEN is a registered trademark of Croda International PLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service