Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

166508

Millipore

SDA - ICR 90mm 20 P

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106217

pH

7.3 ( in water)

Quality Level

Application

Environmental monitoring for isolators & clean rooms, Active or passive air monitoring, Personnel monitoring (fingerprint)

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service