Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

165300

Sigma-Aldrich

L-(+)-2-Amino-4-phosphonobutyric Acid

Synonym(s):

L-(+)-2-Amino-4-phosphonobutyric Acid, L-AP4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H10NO5P
CAS Number:
Molecular Weight:
183.10
MDL number:
UNSPSC Code:
12352200

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

dilute aqueous base: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C4H10NO5P/c5-3(4(6)7)1-2-11(8,9)10/h3H,1-2,5H2,(H,6,7)(H2,8,9,10)/p-2/t3-/m0/s1

InChI key

DDOQBQRIEWHWBT-VKHMYHEASA-L

General description

Phosphonic acid analog of L-glutamic acid. A selective group III metabotropic glutamate receptor agonist. Also a potent, pathway-specific synaptic depressant.

Warning

Toxicity: Irritant (B)

Other Notes

Pin, J.-P., and Duvoisin, R. 1995. Neuropharmacology 34, 1.
Saugstad, J.A., et al. 1995. Mol. Pharmacol. 45, 367.
Tones, M.A., et al. 1995. NeuroReport 7, 117.
Schoepp, D.D. 1994. Neurochem. Int. 24, 439.
Nakanishi, S. 1992. Science 258, 597.
Tanabe, Y., et al. 1992. Neuron 8, 169.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service