Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

152226

Sigma-Aldrich

AEC Substrate Buffer

This AEC Substrate Buffer is validated for use in Immunoblotting and Immunohistochemistry.

Synonym(s):

AEC Substrate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116133

Quality Level

form

liquid

does not contain

preservative

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

shipped in

ambient

storage temp.

2-8°C

General description

AEC Substrate Buffer is a peroxidase substrate buffer supplied as a 10X concentrate (pH 5.0 when diluted). It is used as a diluent for 3-Amino-9-ethylcarbazole (AEC), a widely used chromogen for immunohistochemical staining.

Application

AEC Substrate Buffer has been used in memory B cells enzyme-linked immune absorbent spot (ELISpot) assay.

Warning

Toxicity: Standard Handling (A)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nadine G Rouphael et al.
NPJ vaccines, 6(1), 89-89 (2021-07-16)
In a phase 1 randomized, single-center clinical trial, inactivated influenza virus vaccine delivered through dissolvable microneedle patches (MNPs) was found to be safe and immunogenic. Here, we compare the humoral and cellular immunologic responses in a subset of participants receiving
Hermann O Mayr et al.
Archives of orthopaedic and trauma surgery, 139(3), 383-391 (2019-01-19)
Arthrofibrosis (AF) is the result of increased cell proliferation and synthesis of matrix proteins (collagen I, III, and VI). Especially after invasive knee surgery, e.g., ligament reconstruction or knee replacement, abnormal fibroblast proliferation with pathological periarticular fibrosis can be observed
Evan J Anderson et al.
Vaccines, 8(1) (2020-02-12)
Background: Vaccinia is known to induce antibody and cellular responses. Plasmablast, circulating follicular helper T (cTFH) cells, cytokine-expressing CD4 T cells, and memory B cells were compared between subcutaneous (SC) and needle-free jet injection (JI) recipients of non-replicating modified vaccinia

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service