Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

14-926-M

Sigma-Aldrich

IKKε Protein, active, 10 µg

Active, N-terminal FLAG & GST-tagged, recombinant, human IKKε, full length. For use in Kinase Assays.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.41

biological source

human

Quality Level

recombinant

expressed in baculovirus infected Sf21 cells

mol wt

Mw 109 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

General description

IKBKE is a noncanonical I-kappa-B kinase (IKK) that is essential for regulating antiviral signaling pathways. IKBKE has also been identified as a breast cancer oncogene and is amplified and overexpressed in over 30% of breast carcinomas and breast cancer cell lines. Phosphorylates inhibitors of NF-kappa-B thus leading to the dissociation of the inhibitor/NF-kappa-B complex and ultimately the degradation of the inhibitor. May play a special role in the immune response. Protects cells against DNA damage-induced cell death.
Product Source: N-terminal FLAG and GST-tagged, recombinant, human IKK full length, expressed by baculovirus in Sf21 insect cells.

Application

Research Category
Signaling

Biochem/physiol Actions

Protein Target: IKKe
Target Sub-Family: Misc

Packaging

Also available in 250μg size as catalog # 14-926M and in bulk as catalog # 14-926-K.

Quality

Routinely evaluated by phosphorylation of casein.

Physical form

50mM Tris/HCl pH7.5, 150mM NaCl, 10mM β-glycerophosphate, 1mM sodium orthovanadate, 1mM NaF, 28% v/v glycerol, 0.03% Brij-35, 1mM benzamidine, 0.2mM PMSF, 0.1% 2-mercaptoethanol
Purified by immunoaffinity chromatography.

Storage and Stability

6 months at -70°C

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service