Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

14-918-M

Sigma-Aldrich

JAK1 Protein, active, 10 µg

Active, N-terminal GST-tagged, recombinant, human JAK1, amino acids 866-end, expressed by baculovirus in Sf21 insect cells. Activated in vitro by auto-phosphorylation. For use in Kinase Assays.

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$558.00
250 μG
$4,520.00

$558.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
$558.00
250 μG
$4,520.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

$558.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

human

Quality Level

recombinant

expressed in baculovirus infected Sf21 cells

mol wt

Mw 61 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

Gene Information

human ... JAK1(3716)

General description

Janus kinase 1 (JAK1), is a member of a new class of protein-tyrosine kinases (PTK) characterized by the presence of a second phosphotransferase-related domain immediately N-terminal to the PTK domain. The second phosphotransferase domain bears all the hallmarks of a protein kinase, although its structure differs significantly from that of the PTK and threonine/serine kinase family members. JAK1 is a large, widely expressed membrane-associated phosphoprotein. JAK1 is involved in the interferon-alpha/beta and -gamma signal transduction pathways. The reciprocal interdependence between JAK1 and TYK2 activities in the interferon-alpha pathway, and between JAK1 and JAK2 in the interferon-gamma pathway, may reflect a requirement for these kinases in the correct assembly of interferon receptor complexes. These kinases couple cytokine ligand binding to tyrosine phosphorylation of various known signaling proteins and of a unique family of transcription factors termed the signal transducers and activators of transcription, or STATs.
Product Source: N-terminal GST-tagged, recombinant, human JAK1 amino acids 866-end, expressed by baculovirus in Sf21 insect cells. Activated in vitro by auto-phosphorylation.

Application

Research Category
Signaling

Biochem/physiol Actions

Target Sub-Family: TK

Packaging

Also available in 250μg size as catalog # 14-918M and in bulk as catalog # 14-918-K.

Quality

Routinely evaluated by phosphorylation of peptide GEEPLYWSFPAKKK.

Physical form

50mM Tris/HCl pH7.5, 150mM NaCl, 5mM β-glycerophosphate, 0.5mM sodium orthovanadate, 50% v/v glycerol, 0.02% Triton X-100, 0.5mM EDTA, 2mM DTT. Frozen solution.
Purified using glutathione sepharose.

Storage and Stability

6 months at -70°C

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service