Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

14-912-M

Sigma-Aldrich

TGFBR-1 Protein, active, 10 µg

Active, N-terminal GST-tagged, recombinant, human TGFBR1 amino acids 200–end containing a T204D mutation, expressed by baculovirus in Sf21 insect cells. Purified using glutathione agarose. For use in Kinase Assays.

Synonym(s):

TGF-beta receptor type-1, TGFR-1, Transforming growth factor-beta receptor type I, TGF-beta receptor type I, TbetaR-I, TGF-beta type I receptor, Serine/threonine-protein kinase receptor R4, SKR4, Activin receptor-like kinase 5, ALK-5

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$695.00
250 μG
$5,700.00

$695.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
$695.00
250 μG
$5,700.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

$695.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

human

Quality Level

recombinant

expressed in baculovirus infected Sf21 cells

mol wt

Mw 62 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

Gene Information

human ... TGFBR1(7046)

General description

N-terminal GST-tagged, recombinant, human TGFBR1 amino acids 200–end containing a T204D mutation, expressed by baculovirus in Sf21 insect cells. Purified using glutathione agarose.
Product Source: Expressed by baculovirus in Sf21 insect cells.
Transforming growth factor, beta receptor type I (TGF-beta receptor I) forms a heteromeric complex with TGF-beta receptor type II when bound to TGF-beta. This complex transduces the TGF-beta signal from the cell surface to the cytoplasm. Type II receptors phosphorylate and activate type I receptors which autophosphorylate, then bind and activate SMAD transcriptional regulators. TGF-beta receptor I is found in all tissues, but is most abundant in placenta and least abundant in brain and heart tissue. TGF-beta receptor I is phosphorylated at basal levels in the absence of ligand binding and is multiply phosphorylated, mainly in the GS region. Defects in TGF-beta receptor I are the cause of Loeys-Dietz syndrome type 1A (LDS1A), also known as Furlong syndrome or Loeys-Dietz aortic aneurysm syndrome (LDAS).

Application

Research Category
Inflammation & Immunology

Biochem/physiol Actions

Protein Target: TGFBR1
Target Sub-Family: TKL

Packaging

Also available in 250 µg size --call for pricing and availability and reference catalog number 14-912M when ordering the 250 µg size.

Quality

Routinely evaluated by phosphorylation of casein.

Physical form

50mM Tris-HCl pH7.5, 300mM NaCl, 0.1mM EGTA, 0.03% Brij-35, 270mM sucrose, 1mM benzamidine, 0.2mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution.
Glutathione agarose affinity chromatography

Storage and Stability

Store -70°C. For maximum recovery of product, centrifuge original vial prior to removing the cap.
Rapidly thaw the vial under cold water and immediately place on ice. Aliquot unused material into pre-chilled microcentrifuge tubes and immediately snap-freeze the vials in liquid nitrogen prior to re-storage at -70°C.

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service