Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

14-818-M

Sigma-Aldrich

Met (D1246N) Protein, active, 10 µg

Active, N-terminal 6His-tagged, recombinant, human, Met amino acids 974-end, containing the mutation D1246N, for use in Kinase Assays.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

biological source

human

Quality Level

recombinant

expressed in Sf21 cells

mol wt

Mw 50.1 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

UniProt accession no.

General description

Met D1246N is a germline mutation that has been identified in patients with hereditary papillary renal carcinomas (HPRC). In vitro studies have demonstrated enhanced kinase activity from this mutant.
N-terminal 6His-tagged, recombinant, human, Met amino acids 974-end, containing the mutation D1246N
Product Source: expressed by baculovirus in Sf21 insect cells

Biochem/physiol Actions

Protein Target: Met
Target Sub-Family: TK

Packaging

Available in 250 μg --call for pricing and availability and reference catalog number 14-818M when ordering the 250 μg size.

Physical form

Ni2+/NTA-agarose

Storage and Stability

6 months at -70°C

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service