Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

14-805-M

Sigma-Aldrich

Met (Y1248C) Protein, active, 10 µg

Active, N-terminal 6His-tagged, recombinant, human, Met amino acids 974-end, containing the mutation Y1248C. For use in Kinase Assays.

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$550.00
250 μG
$5,190.00

$550.00


Please contact Customer Service for Availability


Select a Size

Change View
10 μG
$550.00
250 μG
$5,190.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

$550.00


Please contact Customer Service for Availability

biological source

human

Quality Level

recombinant

expressed in baculovirus infected Sf21 cells

mol wt

Mw 50 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

UniProt accession no.

General description

Met Y1248C is a germline mutation that has been identified in patients with hereditary papillary renal carcinomas (HPRC).
N-terminal 6His-tagged, recombinant, human, Met amino acids 974-end, containing the mutation Y1248C.

Packaging

Available in 250 μg --call for pricing and availability and reference catalog number 14-805M when ordering the 250 μg size.

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service