Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

14-752-M

Sigma-Aldrich

GRK7 Protein, active, 10 µg

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

biological source

human

Quality Level

100
300

mol wt

Mw 66 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

General description

N-terminal 6His-tagged, recombinant, full length human GRK7 expressed by baculovirus in Sf21 cells.
Recombinant, full length human GRK7 expressed by baculovirus in Sf21 cells.

Application

Active, N-terminal 6His-tagged, recombinant, full length human GRK7 expressed by baculovirus in Sf21 cells. For use in Kinase Assays.
Research Category
Metabolism
Research Sub Category
Cardiovascular Disease|Metabolic Disorders

Biochem/physiol Actions

  • Protein Target: GRK7
  • Target Sub-Family:AGC

Quality

Routinely evaluated by phosphorylation of 2 mg/mL casein.
Purity was assessed by SDS-PAGE and Coomassie blue staining using 3μg of GRK7.

Unit Definition

Specific Activity: where one unit of GRK7 activity is defined as 1nmol phosphate incorporated into 2mg/ml casein per minute at 30°C with a final ATP concentration of 100μM.

Physical form

10μg of recombinant protein in 100 μl of 50 mM Tris-HCl pH 8.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution.
Format: Purified using Ni2+/NTA agarose. Purity 99% by SDS-PAGE and Coomassie blue staining.

Storage and Stability

Stable for 6 months at -70°C from date of shipment. For maximum recovery of product, centrifuge original vial prior to removing the cap.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service