Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

14-745-M

Sigma-Aldrich

TAO3 Protein, active, 10 µg

Active, N-terminal 6His-tagged, recombinant, human TAO3 amino acids 1-411, N47S. For use in Kinase Assays.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

biological source

human

Quality Level

recombinant

expressed in baculovirus infected Sf21 cells

Assay

84% (Coomassie blue staining, SDS-PAGE)

specific activity

915 U/mg

mol wt

Mw 50.2 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

Gene Information

human ... TAOK3(51347)

General description

N-terminal 6His-tagged, recombinant, human TAO3 amino acids 1-411; N47S.
Product Source: Recombinant, human TAO3 amino acids 1-411; N47S. Expressed by baculovirus in Sf21 cells.

Application

Research Category
Metabolism
Research Sub Category
Cardiovascular Disease

Metabolic Disorders

Biochem/physiol Actions

Protein Target: TAO3
Target Sub-Family: STE

Quality

Routinely evaluated by phosphorylation of 0.8 mg/mL myelin basic protein.
Purity was assessed by SDS-PAGE and Coomassie blue staining using 3μg of TAO3.

Unit Definition

Specific Activity: where one unit of TAO3 activity is defined as 1nmol phosphate incorporated into 0.8mg/ml myelin basic protein per minute at 30°C with a final ATP concentration of 100μM.

Physical form

10 μg of enzyme in 6.3 μL of 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij 35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution.
Ni2+/NTA-agarose

Storage and Stability

Stable for 6 months at -70°C from date of shipment. For maximum recovery of product, centrifuge original vial prior to removing the cap.
Rapidly thaw the vial under cold water and immediately place on ice. Aliquot unused material into pre-chilled microcentrifuge tubes and immediately snap-freeze the vials in liquid nitrogen prior to re-storage at -70°C.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service