Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

14-721-M

Sigma-Aldrich

EGFR (T790M, L858R) Protein, active, 10 µg

Active, N-terminal GST-tagged, recombinant human EGFR, amino acids 696-end containing the mutations T790M & L858R, for use in Enzyme Assays.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

biological source

human

Quality Level

recombinant

expressed in baculovirus infected Sf21 cells

product line

Upstate®

specific activity

(For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.)

mol wt

Mw 85.8 kDa

purified by

(Glutathione-agarose)

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable

accession no.

 (FUNCTION: SwissProt: P00533 # Isoform 2/truncated isoform may act as an antagonist. )
NM_005228.3

UniProt accession no.

relevant disease(s)

arthritis; obesity; osteoporosis; metabolic disorders

General description

N-terminal GST-tagged, recombinant human EGFR, amino acids 696end containing the mutations T790M and L858R

Application

Research Category
Metabolism

Inflammation & Immunology
Research Sub Category
Obesity

Metabolic Disorders

Osteoporosis

Arthritis

Biochem/physiol Actions

Protein Target: EGFR
Target Sub-Family: TK

Packaging

Also available in 10 μg size and in bulk

Quality

routinely evaluated by phosphorylation of 250μM substrate peptide (GGMEDIYFEFMGGKKK)

Target description

EGFR

Physical form

Glutathione-agarose

Storage and Stability

6 months at -70°C from date of shipment

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service