Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

14-692-M

Sigma-Aldrich

DAPK1 Protein, active, 10 µg

Active, N-terminal GST-tagged, recombinant, human DAPK1, amino acids 1-296, for use in Kinase Assays.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

biological source

human

Quality Level

100
300

recombinant

expressed in baculovirus infected Sf21 cells

mol wt

Mw 60.9 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

Gene Information

human ... DAPK1(1612)

General description

N-terminal GST-tagged, recombinant, human DAPK1, amino acids 1-296

Biochem/physiol Actions

Protein Target: DAPK1

Quality

routinely evaluated by phosphorylation of ZIPtide (Catalog #12-545)

Storage and Stability

stable 6 months at -70°C from date of shipment

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

DAP-kinase as a target for drug design in cancer and diseases associated with accelerated cell death.
Bialik, Shani and Kimchi, Adi
Seminars in cancer biology, 14, 283-294 (2004)
Epigenetic change in pituitary tumorigenesis.
Farrell, W E and Clayton, R N
Endocrine-Related Cancer, 10, 323-330 (2003)
Anastasia V Velentza et al.
Pharmacology & therapeutics, 93(2-3), 217-224 (2002-08-23)
Death-associated protein kinase (DAPK) is a calmodulin-regulated serine/threonine protein kinase associated with neuronal cell death in animal models of disease. The recent determination of the 1.5A crystal structure of the catalytic kinase domain of DAPK, the discovery of amino acid
The DAP-kinase family of proteins: study of a novel group of calcium-regulated death-promoting kinases.
Shohat, Galit, et al.
Biochimica et Biophysica Acta, 1600, 45-50 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service