Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.46764

Millipore

Plate Count Agar Kit

Ready-to-Use, 55mm plates with EZPAK 0.45 White Gridded Membrane for the enumeration of total viable count using the Membrane Filtration method, released through an ISO 17025 accredited lab.

Synonym(s):

Tryptone Glucose Yeast Agar, Casein-Peptone Dextrose Yeast Extract Agar, Standard Methods Agar, PCA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106217
NACRES:
NA.74

material

gridded white mixed cellulose esters (MCE) membrane

Quality Level

Agency

ISO

form

plate (ready-to-use)

shelf life

9 mo.

feature

ready-to-use

manufacturer/tradename

ReadyPlate

packaging

pkg of (150x55mm Plates and 150 membranes)

technique(s)

cell counting: suitable (total viable count (TVC))

pore size

0.45 μm pore size

application(s)

filtration
food and beverages
microbiology

storage temp.

15-25°C

suitability

aerobic bacteria (total count)

General description

Plate Count Agar Kit contains
ReadyPlate PC Agar plates of 55mm diameter is a selective culture medium. The pH of the medium is in the optimum range of 6.8-7.2 for the growth of bacteria. The media is relatively rich in its nutrients and does not contain any inhibitors or indicators. The enzymatic digest of casein serves as a nitrogen source with high levels of free amino acids followed by yeast extract, which is a rich source of vitamins. The carbon source for the growth of bacteria is served by glucose thereby providing efficient growth of bacteria.
EZ-Pak Filters are ready-to-use, sterile filters made of Mixed cellulose esters (MCE) with 0.45 μm pore size, white, gridded and 50mm diameter. These filters possess high recoveries.

Application

Plate Count Agar Kit contains 55mm plates with EZPAK 0.45 μm white gridded membrane for the simultaneous enumeration of total viable count using the Membrane Filtration method in water samples

Legal Information

READYPLATE is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service