Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.46752

Millipore

Tryptic Soy Broth- Ready-to-use Bags, Tubes & Ampoules

closure type, with KPC connector, box of 22 × 10 L

Synonym(s):

CASO, CASO Broth, Casein Soya Broth, TSB, Tryptic Soy Broth, Tryptone Soy Broth, Tryptic Soy Broth

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
Pricing and availability is not currently available.

Agency

EP 2.6.1
JP 4.06
USP 71

Quality Level

sterility

sterile; γ-irradiated
sterile; autoclaved

form

liquid

feature

closure type with KPC connector
ready-to-use

packaging

box of 22 × 10 L

bag volume

10 L

capacity

22x10 L

pH

7.3

application(s)

pharmaceutical
sterility testing

storage temp.

2-25°C

suitability

nonselective for

Looking for similar products? Visit Product Comparison Guide

General description

Ready-to-use and sterile TSB, contained in a 10L self-collapsing bag, with an KPC connector. The bag is tripple-bagged and irradiated. Tryptic Soy Broth is a universal complex medium for the isolation and cultivation of microorganisms, especially for the microbiological aseptic process simulation (Media Fill Test). The broth is suitable for media fill tests according to the recommendations of the "FDA Aseptic Guide" in aseptic production lines. The formulation of the basic medium (Soybean-Casein Digest Broth) is prepared according to the recommendations of the current European, and United States Pharmacopoeia.

Application

Tryptic Soy Broth irradiated is suitable for use in media fill tests or aspetc process simulation. The included peptones and glucose allow the cultivation and isolation of a broad range of fastidious aerobic bacteria, yeasts and molds.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service