Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

14-674-M

Sigma-Aldrich

EphB1 Protein, active, 250 µg

Sign Into View Organizational & Contract Pricing

Select a Size

250 μG
$5,190.00

$5,190.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 μG
$5,190.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

$5,190.00


Please contact Customer Service for Availability

Request a Bulk Order

recombinant

expressed in baculovirus infected Sf21 cells

Quality Level

description

protein target EphB1

purified by

(Ni2+/NTA-agarose)

technique(s)

activity assay: suitable

UniProt accession no.

Gene Information

human ... EPHB1(2047)

General description

FUNCTION: SwissProt: P54762 # Receptor for members of the ephrin-B family. Binds to ephrin-B1, -B2 and -B3. May be involved in cell-cell interactions in the nervous system.
SIZE: 984 amino acids; 109885 Da
SUBUNIT: The ligand-activated form interacts with GRB2, GRB10 and NCK through their respective SH2 domains. The GRB10 SH2 domain binds EPHB1 through Tyr-928, while GRB2 binds residues within the catalytic domain. The NCK SH2 domain binds EPHB1 through Tyr-594. Interacts with PRKCABP (By similarity).
SUBCELLULAR LOCATION: Membrane; Single-pass type I membrane protein.
TISSUE SPECIFICITY: Preferentially expressed in brain.
SIMILARITY: SwissProt: P54762 ## Belongs to the protein kinase superfamily. Tyr protein kinase family. Ephrin receptor subfamily. & Contains 2 fibronectin type-III domains. & Contains 1 protein kinase domain. & Contains 1 SAM (sterile alpha motif) domain.

Packaging

Also available in 10 μg size (cat#14-674) and in bulk (cat# 14-674-K).

Quality

routinely evaluated by phosphorylation of 250μM Src Substrate Peptide (Catalog # 12-140

Physical properties

51.4kDa

Storage and Stability

6 months at -70°C from date of shipment

Analysis Note

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service