Skip to Content
MilliporeSigma
All Photos(2)

Documents

1.46749

Millipore

Veg. Peptone Broth 10l Lynx 1PC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NA.74

Agency

EP
JP
USP

Quality Level

sterility

sterile; sterile-filtered

form

liquid

feature

ready-to-use

packaging

pkg of 1 pieces

technique(s)

media fill: suitable

volume

10 L
10000 mL

pH

7.3

application(s)

aseptic process simulation
pharmaceutical

compatibility

stand alone

storage temp.

2-25°C

suitability

nonselective for

General description

Ready-to-use 10L bag with Lynx connector, tripple bagged and irradiated. Vegetable Peptone Broth (or Tryptic Sor Broth, non-animal origin) is a universal complex medium for the isolation and cultivation of microorganisms, especially for the Aseptic process simulation (Media Fill Test). The broth is suitable for media fill tests according to the recommendations of the “FDA Aseptic Guide” in aseptic production lines. The exclusive use of raw materials of non-animal origin allows the elimination of a potential contamination risk with TSE/BSE. The formulation of the basic medium (Vegetable Peptone Broth) is prepared according to the recommendations of the current European and United States Pharmacopoeia.

Application

Vegetable Peptone broth (VPB) is suitable for use in media fill tests. Growth promotion studies show very good results compared to conventional tryptic soy broth (TSB)

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service