Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

14-670-M

Sigma-Aldrich

TSSK1 Protein, active, 10 µg

Sign Into View Organizational & Contract Pricing

Select a Size

250 μG
$4,380.00

$4,380.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 μG
$4,380.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

$4,380.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

recombinant

expressed in baculovirus infected Sf21 cells

description

protein target TSSK1

purified by

(Ni2+/NTA-agarose)

technique(s)

activity assay: suitable

UniProt accession no.

Gene Information

human ... TSSK1B(83942)

General description

FUNCTION: SwissProt: Q9BXA7 # May be involved in a signaling pathway during male germ cell development or mature sperm function (By similarity).
COFACTOR: Magnesium.
SIZE: 367 amino acids; 41618 Da
PTM: Autophosphorylated.
SIMILARITY: SwissProt: Q9BXA7 ## Belongs to the protein kinase superfamily. Ser/Thr protein kinase family. & Contains 1 protein kinase domain.

Packaging

Also available in 10 μg size (cat#14-670) and in bulk (cat#14-670-K).

Quality

Routinely evaluated by phosphorylation of CHKtide (Catalog # 12-373)
routinely evaluated by phosphorylation of CHKtide (Catalog # 12-373)

Physical properties

46kDa

Storage and Stability

6 months at -70°C

Analysis Note

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service