Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

1.46697

Millipore

Tryptic Soy Agar - Ready-to-use Contact Plates

Neutralizer A, ICR plus with lockable lid, plate diam. 55 mm, sterile; γ-irradiated, pkg of 10 plates Triple packed, suitable for surface monitoring

Synonym(s):

CASO agar, Casein Soybean Digest Agar with neutralizer, TSA, TSA with Neutralizer A

Sign Into View Organizational & Contract Pricing

Select a Size

20 PKG
$118.00
200 PKG
$1,060.00

$118.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
20 PKG
$118.00
200 PKG
$1,060.00

About This Item

UNSPSC Code:
41106231
NACRES:
NA.74

$118.00


Please contact Customer Service for Availability

Request more information

material

polystyrene plate

Quality Level

Agency

EP
USP

sterility

sterile; γ-irradiated

form

plate (ready-to-use; with safety lock)

feature

ready-to-use

composition

Neutralizer A

packaging

pkg of 10 plates Triple packed
box of 20 ea
box of 200 ea

storage condition

protect from light (Tightly closed. Dry)

plate diam.

55 mm

color

yellow to yellow-orange

application(s)

pharmaceutical
surface monitoring

compatibility

stand alone

storage temp.

15-25°C

suitability

bacteria
fungi

Looking for similar products? Visit Product Comparison Guide

General description

Neutralizer A ICR+ is designed for the determination of the total microbial aerobic and anaerobic count on dry, sanitized surfaces and personnel in Clean rooms and Isolators. It is used for the cultivation and isolation of a wide range of bacteria, yeasts and molds. Neutralizer A plate contains a mixture of neutralizers which inactivates potential residues of disinfectants containing active agents such as Alcohol (ethanol or isopropyl alcohol), Aldehyde, Glucoprotamine, Hydrogen Peroxide, Peracetic acid, Phenols and quaternary ammonium compounds and Polyhexamethylene biguanides. In addition, the ICRplus (plus or “+”) plates are supplied with a lockable lid for safe transport after sampling without the risk of losing the lid as well as for aerobic (close position) and anaerobic incubation (vent position).

Application

Neutralizer A plate is used for cultivation and isolation of a wide range of bacteria, yeasts and molds. The media finds its application in environmental monitoring by determining the total microbial aerobic and anaerobic count on dry, sanitized surfaces and personnel in Isolators and Clean Rooms

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Microbial monitoring; environmental monitoring; interim storage; ICR contact plates; contact plates; TSA w. LTHThio Contact ICR+

Related Content

Contact Plates and Swabs for Personnel and Surface Monitoring in Cleanrooms and Isolators

Questions

  1. Referring to product 1.46697 (.0020 and 0200) with neutralizer A: what is the exact composition of neutralizer A?

    1 answer
    1. The composition of the neutralizer A used in the formulation of 1466970020 and 1466970120 is proprietary and cannot be disclosed. While the composition cannot be disclosed, this mixture neutralizes a broad range of disinfectants such as hydrogen peroxide, sodium hypochlorite, alcohols, quaternary ammonium compounds. The full neutralizer inactivation capabilities of this neutralizer A can be reviewed in our validation report, available on demand.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service