1.46577
Sabouraud Dextrose Agar
lid, ICR: non lockable plate, plate diam. 90 mm, sterile; irradiated, triple packed of 20 or 120 plates, suitable for surface monitoring
Synonym(s):
ICR plate, SDA
Select a Size
$93.50
Select a Size
About This Item
$93.50
Recommended Products
material
polystyrene plate
Quality Level
Agency
EP
USP
sterility
sterile; irradiated
form
plate (ready-to-use)
feature
lid ICR: non lockable plate
ready-to-use
packaging
triple packed of 20 or 120 plates
technique(s)
active air sampling: suitable
passive air sampling: suitable
plate diam.
90 mm
application(s)
pharmaceutical
surface monitoring
compatibility
stand alone
storage temp.
2-25°C
suitability
molds
yeasts
Looking for similar products? Visit Product Comparison Guide
General description
Packaging: 90 mm settle plate (20 or 120 plates per box)
Synonym: SDA, Sabouraud Dextrose Agar
Application
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service