Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.46509

Millipore

Transparent, gamma irradiated Bags

box of 125 bags, polyethylene, sterile; irradiated

Synonym(s):

Sterile transparent bags

Sign Into View Organizational & Contract Pricing

Select a Size

125 EA
$656.00

$656.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
125 EA
$656.00

About This Item

UNSPSC Code:
41122409
NACRES:
NC.81

$656.00


Please contact Customer Service for Availability

Request more information

material

polyethylene sleeve

Quality Level

sterility

sterile; irradiated

feature

ready-to-use

packaging

box of 125 bags

bag L × W

21 cm × 21 cm

application(s)

microbiology
pharmaceutical

compatibility

for use with Contact Plate

storage temp.

2-25°C

General description

Transparent Bags are sterile bags used to protect the plates from secondary contamination during transport outside of the cleanroom zone. These polyethylene bags are irradiated and can be used for a safe transport of Contact & Settle plates from production site to microbiology laboratory.

Application

Transparent Bags are used to protect the plates from secondary contamination during transport

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Settle plates for isolators and critical cleanrooms

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service