Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.46457

Millipore

Tryptic Soy Agar - Ready-to-use Bottles & Tubes

bottle capacity 250 mL, bottle filling volume 200 mL, closure type, blue screw cap with 3 loci, box of 6 bottles

Synonym(s):

CASO Agar, Casein Soybean Digest Agar, Tryptone Soya Agar, TSA, Tryptic Soy Agar EP+USP

Sign Into View Organizational & Contract Pricing

Select a Size

6 X 200 ML
$103.00

$103.00


Please contact Customer Service for Availability


Select a Size

Change View
6 X 200 ML
$103.00

About This Item

UNSPSC Code:
41106200
NACRES:
NA.74

$103.00


Please contact Customer Service for Availability

Agency

EP
USP

Quality Level

description

For cultivation & isolation of fastidious bacteria, yeasts, and fungi

sterility

sterile

form

liquid

shelf life

12 mo.

feature

closure type blue screw cap with 3 loci
ready-to-use

packaging

box of 6 bottles
single packed of (200 ml in 250 ml bottle with blue screw cap and 3 loci (6 bottles per box))

technique(s)

direct inoculation: suitable
sterile filtration: suitable

bottle capacity

250 mL

bottle filling volume

200 mL

pH

7.3

application(s)

cosmetics
food and beverages
microbiology
pharmaceutical
sterility testing

storage temp.

2-25°C

Looking for similar products? Visit Product Comparison Guide

General description

Tryptic Soy Agar is a complex growth medium. It is suitable for a large range of applications, including microbial cultivation, maintenance of stock culture, and total aerobic microbial count. This medium contains peptones, amino acids, and polypeptides to ensure the growth of a variety of microorganisms. Sodium chloride maintains osmotic equilibrium in the medium.

Application

Tryptic Soy Agar (TSA) is suitable for the cultivation and isolation of fastidious bacteria, yeasts, and fungi. TSA is especially used to determine bacterial contamination in cosmetics, drinking water, and wastewater.

Features and Benefits

  • Ready-to-use media
  • Bottles with screw caps
  • Suitable for pour plate method

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service