Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.46364

Millipore

ReadyTube® MRS Medium

For the isolation, enumeration and cultivation of Lactobacillus sp. according to ISO 15214

Synonym(s):

ReadyTube® MRS Medium Agar according to de Man, Rogosa and Sharpe"

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106225
NACRES:
NA.74
Pricing and availability is not currently available.

Agency

ISO 15214

Quality Level

form

solid (medium)

feature

ready-to-use

manufacturer/tradename

ReadyTube®

packaging

pkg of (200 ml in 250 ml bottle with blue screw cap and 3 loci (6 bottles per box))

pH

5.6-5.9 ( in H2O)

application(s)

food and beverages
industrial qc
microbiology

storage temp.

2-25°C

suitability

lactic acid bacteria

General description

ReadyTube® MRS Medium is based on the formulation of deMan, Rogosa and Sharpe used for the isolation enumeration and detection of Lactobacillus sp. The enzymatic Digest of Casein supply nitrogenous and carbonaceous compounds. Yeast extract provides vitamin B complex and glucose is the fermentable carbohydrate and energy source. Polysorbate 80 supplies fatty acids required for the metabolism of Lactobacilli. Sodium acetate inhibits the growth of Streptococci, molds and many other microorganisms. Magnesium sulphate and manganese sulphate provide essential ions for multiplication of Lactobacilli. Phosphates provide good buffering action in the media.

Application

ReadyTube® MRS Medium is used for the isolation, enumeration and cultivation of Lactobacillus sp. from all types of materials

Legal Information

READYTUBE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service