Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.46327

Millipore

NaCl Peptone Broth

ready-to-use, tube volume 9 mL , filling volume

Synonym(s):

MRD, Maximum Recovery Diluent, Saline Peptone Solution, Saline Peptone Water, Sodium Chloride Peptone Solution, Sodium Chloride Peptone Water

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106229
NACRES:
NA.74

description

Diluent for maximum recovery of microorganisms

Quality Level

form

liquid (ready-to-use)

shelf life

12 mo.

feature

closure type screw cap with septum
ready-to-use

packaging

single packed of (9 ml in 17 ml tube with blue cap (100 tubes per box))
tube of

technique(s)

direct inoculation: suitable
sterile filtration: suitable

tube capacity

17 mL

tube volume

9 mL , filling volume

color

clear colorless

pH

7

application(s)

cosmetics
food and beverages
microbiology
pharmaceutical

storage temp.

2-25°C

General description

NaCl Peptone Broth is a general diluent used for sample preparation. The solution′s low peptone content maintains organisms′ viability without promoting growth. Sodium chloride maintains the osmotic balance of the medium, thereby preserving cell integrity. The neutral range of medium pH is optimum for the viability of microorganisms.

Application

NaCl Peptone Broth is used as a protective and isotonic diluent for different test method in microbiological area and for maximum recovery of microorganisms.

Features and Benefits

  • Our ready-to-use media provide the highest level of quality and testing confidence. They have been formulated and tested to meet the pharmacopoeia requirements.
  • Sterility testing media and rinse solutions are manufactured in an ISO 9001, environmentally controlled production center. Each lot undergoes a stringent quality control (QC) procedure, including pH, sterility, and growth promotion tests.
  • Our manufacturing approach ensures the highest level of clarity for our media and rinsing fluids, therefore improving accuracy, and significantly reducing the risk of incorrect interpretation and false results.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service