Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

1.46303

Millipore

Sabouraud Dextrose Agar

tube capacity × filling volume 25 mL × 18 mL, closure type, yellow cap, box of 20 and 100 tubes

Synonym(s):

SDA, SDA Ready-to-use media

Sign Into View Organizational & Contract Pricing

Select a Size

100 PKG
$331.00

$331.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
100 PKG
$331.00

About This Item

UNSPSC Code:
41106204
NACRES:
NA.74

$331.00


Please contact Customer Service for Availability

Request more information

Agency

EP 2.6.13
JP 4.05
USP 62

Quality Level

description

For microbial examination of non-sterile pharmaceutical products

sterility

sterile

form

gelatinous solid

shelf life

12 mo.

feature

closure type yellow cap
ready-to-use

packaging

box of 20 and 100 tubes
tube of

technique(s)

direct inoculation: suitable
sterile filtration: suitable

tube capacity × filling volume

25 mL × 18 mL

color

clear yellowish

pH

5.6

application(s)

cosmetics
food and beverages
microbiology
pharmaceutical

storage temp.

2-25°C

suitability

molds
yeasts

General description

Sabouraud Dextrose Agar is a complex medium for the cultivation and isolation of yeasts and molds. The high concentration of Dextrose and the low pH promote the growth of yeast and molds while inhibiting bacterial growth. The media also promotes the formation of spores (conidia and sporangia) and pigments of yeasts and molds.

Application

Sabouraud Dextrose Agar (SDA) is designed for the determination of the total count of yeasts and molds. The media is suitable for the microbiological examination of non-sterile products.

Features and Benefits

  • Our ready-to-use media provide the highest level of quality and testing confidence. They have been formulated and tested to meet the pharmacopoeia requirements.
  • Sterility testing media and rinse solutions are manufactured in an ISO 9001, environmentally controlled production center. Each lot undergoes a stringent quality control (QC) procedure, including pH, sterility, and growth promotion tests.
  • Our manufacturing approach ensures the highest level of clarity for our media and rinsing fluids, therefore improving accuracy, and significantly reducing the risk of incorrect interpretation and false results.

Storage and Stability

  • The product can be used for sampling until the expiry date if stored upright, protected from light, andproperly sealed at +2 °C to +25 °.
  • The testing procedures described on the CoA can be started up to the expiry date printed on thelabel.

Other Notes

Agar media should not be melted a second time after solidifying

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service