Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.46206

Millipore

Tryptic Soy Broth- Ready-to-use Bags, Tubes & Ampoules

tube capacity × filling volume 25 mL × 15 mL, closure type, blue screw cap with 3 loci, box of 100 tubes

Synonym(s):

CASO, CASO Broth, Casein Soya Broth, TSB, Tryptic Soy Broth, Tryptone Soy Broth

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106214

Agency

EP
JP
USP

Quality Level

sterility

sterile; autoclaved

form

liquid

feature

closure type blue screw cap with 3 loci
ready-to-use

packaging

box of 100 tubes

tube capacity × filling volume

25 mL × 15 mL

color

clear gold Media

application(s)

pharmaceutical
sterility testing

Looking for similar products? Visit Product Comparison Guide

General description

TSB 15 ml in 25 ml tube.
Tryptic Soy Broth is a universal complex medium. It supports the growth of a wide variety of aerobic and facultative anaerobic microorganisms, including fungi by the content of two highly nutritious peptones.

Application

Tryptic Soy Broth is used for the isolation and cultivation of fastidious aerobic bacteria, yeasts and molds. The medium is suitable for sterility testing of non-sterile substances, preparations, and products. It is also used as pre-enrichment broth for the test for specified microorganisms or total viable aerobic count determinations using the MPN method.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service