Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.46142

Millipore

Buffered Peptone Water

ReadyTube®, according to ISO 6887, FDA BAM, EP, bottle capacity 17 mL, bottle filling volume 9 mL, closure type, white screw cap, box of 20 tubes

Synonym(s):

Alkaline peptone water, BPW, ReadyTube® BPW, Tryptone Phosphate Water

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NA.74

Agency

EP
according to ISO 6887

reg. compliance

FDA BAM

form

liquid (ready-to-use)
liquid

feature

closure type white screw cap
ready-to-use

packaging

box of 20 tubes

manufacturer/tradename

ReadyTube®

bottle capacity

17 mL

bottle filling volume

9 mL

pH

7.0 ( in H2O, ready-to-use solution)

application(s)

cosmetics
environmental
food and beverages
microbiology

storage temp.

2-25°C

suitability

Salmonella spp.

General description

ReadyTube® is ready to use media to provide the highest level of quality and testing confidence for improving accuracy and significantly reducing the risk of false results. ReadyTube® buffered peptone water is pre-enrichment medium, free from inhibitors and is well buffered. It provides conditions for the resuscitation of the cells that have been injured by processes of food preservation. The media contains Peptone, including an enzymatic digest of casein, as a source of carbon, nitrogen, vitamins, and minerals. Sodium chloride maintains the osmotic balance, and phosphates buffer the medium. The broth is rich in nutrients enabling it to produce high resuscitation rates for sub-lethally injured bacteria and support intense growth.

Application

ReadyTube® Buffered peptone water is used as a diluent to detect bacteria, particularly pathogenic Enterobacteriaceae such as Salmonella and Cronobacter, from food, animal feed, and water, and as a diluent for sample preparation.

Legal Information

READYTUBE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service