Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.46049

Millipore

HEIMPLATE R2A Agar chromogenic - LI

ready-to-use, 90 mm settle plates, suitable for microbiology

Synonym(s):

Reasoner´s 2A agar chromogenic for long incubation

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106217
NACRES:
NA.74

Quality Level

sterility

sterile

form

plate (ready-to-use)

feature

ready-to-use

packaging

pack of 20 units

technique(s)

microbiological culture: suitable

pH

7.2 ( in H2O)

application(s)

microbiology
pharmaceutical

storage temp.

15-25°C

suitability

heterotrophic bacteria

General description

R2A Agar (Reasoner´s 2A agar) is designed to promote the growth of aerobic, heterotrophic microorganisms that are adapted to low nutrient environments such as water. The low amount of nutrients in the agar allows the growth of aerobic, heterotrophic microorganisms, which are adapted to low nutrient content. The Gram-negative, Gram-positive bacteria, and several yeast colonies appear red due to the chromogenic agent, which facilitates the evaluation of the plates. However, molds are not affected by the chromogenic agent.

Application

R2A Agar chromogenic - LI in 90 mm settle plates is designed for the determination of the total microbial count in water for injections in bulk, highly purified water, and purified water in bulk.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service