Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.46046

Millipore

BPLS Agar - LI 30ml

Application: Isolation and counting of Salmonella and Shigella
Packaging: 90 mm settle plate with 30 ml filling volume (20 plates/box) for long incubation (LI)
Synonym: Brilliant-Green Phenol-Red Lactose Sucrose Agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200

sterility

sterile

form

plate (ready-to-use)

feature

ready-to-use

technique(s)

microbiological culture: suitable

pH

6.9 ( in H2O)

application(s)

microbiology
pharmaceutical

suitability

Salmonella spp.
Shigella spp.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service